(2-Fluorophenyl)acetone ≥97.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
F0970-5G F0970-25G
TCIAF0970-5GEA 71.5 EUR
TCIAF0970-5G TCIAF0970-25G
(2-Fluorophenyl)acetone ≥97.0% (by GC)
(2-Fluorophenyl)acetone
Formule: C₉H₉FO
Kookpunt: 47°C
Vlampunt: 85°C
MDL Nummer: MFCD00000325
CAS nummer: 2836-82-0
EINECS: 220-627-5
Order Now
Specification Test Results
Appearance White to Yellow to Green clear liquid
Purity (GC) min. 97.0 %