3-Fluoro-2-formylpyridine ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
F0975-200MG F0975-1G
TCIAF0975-200MGEA 20.9 EUR
TCIAF0975-200MG TCIAF0975-1G
3-Fluoro-2-formylpyridine ≥98.0% (by GC)
3-Fluoro-2-formylpyridine
Formule: C₆H₄FNO
Kookpunt: 112°C
MDL Nummer: MFCD07781234
CAS nummer: 31224-43-8
Order Now
Specification Test Results
Appearance White to Green to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 50.0 to 54.0 deg-C