2-Fluoro-5-nitropyridine ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
F0983-5G F0983-25G
TCIAF0983-5GEA 27.5 EUR
TCIAF0983-5G TCIAF0983-25G
2-Fluoro-5-nitropyridine ≥98.0% (by GC)
2-Fluoro-5-nitropyridine
Formule: C₅H₃FN₂O₂
Kookpunt: 87°C
Smeltpunt: 18°C
MDL Nummer: MFCD03095059
CAS nummer: 456-24-6
EINECS: 207-260-6
Order Now
Specification Test Results
Appearance White or Colorless to Yellow powder to lump to clear liquid
Purity (GC) min. 98.0 %