Fmoc-N-methyl-L-leucine ≥98.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: N-Fmoc-N-methyl-L-leucine, Fmoc-N-Me-Leu-OH, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N-methyl-L-leucine

F1028-1G F1028-5G
TCIAF1028-1GEA 31.9 EUR
TCIAF1028-1G TCIAF1028-5G
Fmoc-N-methyl-L-leucine ≥98.0% (by HPLC, titration analysis)
Fmoc-N-methyl-L-leucine
Formule: C₂₂H₂₅NO₄
MW: 367.445 g/mol
MDL Nummer: MFCD00151933
CAS nummer: 103478-62-2

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) gt. 98.0 %
Melting point 111.0 to 115.0 deg-C
Specific rotation [a]20/D -19.0 to -22.0 deg (C=1,DMF)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR