6-Fluoroquinoline ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
F1034-1G F1034-5G
TCIAF1034-1GEA 48.4 EUR
TCIAF1034-1G TCIAF1034-5G
6-Fluoroquinoline ≥98.0% (by GC)
6-Fluoroquinoline
Formule: C₉H₆FN
Kookpunt: 126°C
MDL Nummer: MFCD01685512
CAS nummer: 396-30-5
Order Now
Specification Test Results
Appearance Light yellow to Brown clear liquid
Purity (GC) min. 98.0 %