3-Fluorobenzenesulphonamide ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
F1141-1G
F1141-5G
TCIAF1141-1GEA
44
EUR
TCIAF1141-1G
TCIAF1141-5G
3-Fluorobenzenesulphonamide ≥98.0% (by GC)
3-Fluorobenzenesulphonamide
Formule:
C₆H₆FNO₂S |
MDL Nummer:
MFCD00042284 CAS nummer: 1524-40-9 UN: 2811 ADR: 6.1,III |
|
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 128.0 to 132.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...