Fraxinellone ≥96.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

F1187-10MG F1187-50MG
TCIAF1187-10MGEA 119 EUR
TCIAF1187-10MG TCIAF1187-50MG
Fraxinellone ≥96.0% (by GC)
Fraxinellone
Formule: C₁₄H₁₆O₃
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD11101451
CAS nummer: 28808-62-0
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 96.0 %
Melting point 114.0 to 118.0 deg-C
Specific rotation [a]20/D -37.0 to -41.0 deg (C=1, CHCL3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR