1-Fluoro-2-iodoethane ≥98.0% (by GC) stabilised
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Stabilisatie: Stabilised with copper chip
F1190-1G F1190-5G
TCIAF1190-1GEA 24.2 EUR
TCIAF1190-1G TCIAF1190-5G
1-Fluoro-2-iodoethane ≥98.0% (by GC) stabilised
1-Fluoro-2-iodoethane
Formule: C₂H₄FI
Kookpunt: 102°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00748131
CAS nummer: 762-51-6
UN: 1993
ADR: 3,II
Order Now
Specification Test Results
Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %