4-Fluorophenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: 2-(4-Fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-Fluorophenylboronic acid Pinacol ester, 1-Fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene

F1201-25G F1201-5G
TCIAF1201-25GEA 151 EUR
TCIAF1201-25G TCIAF1201-5G
4-Fluorophenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
4-Fluorophenylboronic acid pinacol ester
Formule: C₁₂H₁₆BFO₂
MDL Nummer: MFCD03453665
CAS nummer: 214360-58-4

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR