Fmoc-Tle-OH ≥98.0% (by HPLC, titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-tert-leucine, (S)-2-(Fmoc-amino)-3,3-dimethylbutyric acid, Fmoc-Tle-OH, Fmoc-α-t-butylglycine, (S)-2-[[[(9H-Fluoren-9-yl)methoxy]carbonyl]amino]-3,3-dimethylbutanoic acid, N-Fmoc-L-tert-leucine, Fmoc-tBu-Gly-OH, (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3,3-dimethylbutanoic acid
F1251-5G
F1251-25G
TCIAF1251-5GEA
64.9
EUR
TCIAF1251-5G
TCIAF1251-25G
Fmoc-Tle-OH ≥98.0% (by HPLC, titration analysis)
Fmoc-Tle-OH
Formule:
C₂₁H₂₃NO₄ MW: 353.418 g/mol |
MDL Nummer:
MFCD00065649 CAS nummer: 132684-60-7 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 123.0 to 127.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...