Fmoc-Lys(Dde)-OH

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: Nalpha-[(9H-Fluoren-9-ylmethyloxy)carbonyl]-Nepsilon-1-(4,4-dimethyl-2,6-dioxocyclohex-1-ylidene)ethyl-L-lysine, Fmoc-Lys(Dde)-OH, Fmoc-L-Lys(Dde)-OH, Nalpha-Fmoc-Nepsilon-1-(4,4-dimethyl-2,6-dioxocyclohex-1-ylidene)ethyl-L-lysine

F1270-1G F1270-5G
TCIAF1270-1GEA 119 EUR
TCIAF1270-1G TCIAF1270-5G
Fmoc-Lys(Dde)-OH
Fmoc-Lys(Dde)-OH
Formule: C₃₁H₃₆N₂O₆
MW: 532.637 g/mol
MDL Nummer: MFCD00278818
CAS nummer: 150629-67-7

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 130.0 to 134.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR