1-Pentanesulphonic acid sodium salt

Leverancier: Serva
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: Sodium 1-pentanesulphonate, Sodium pentane-1-sulphonate, Pentane-1-sulphonic acid sodium salt, Sodium pentanesulphonate, 98%, Sodium pentanesulphonate

31812.01 31812.02
SERA31812.01EA 0 EUR
SERA31812.01 SERA31812.02
1-Pentanesulphonic acid sodium salt
1-Pentanesulphonic acid sodium salt
Formule: C₅H₁₁O₃SNa
Smeltpunt: > 300 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00007541
CAS nummer: 22767-49-3
EINECS: 245-208-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ddb1c23c8a6fe3c156a3aacb62721e1d1a34efb0&vid=1936dcdb47ba374ce21fd758660f7ca6ba95d254&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR