Tripropionin ≥95.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
Glycerol Tripropionate
G0090-25G
G0090-500G
TCIAG0090-25GEA
17.6
EUR
TCIAG0090-25G
TCIAG0090-500G
Tripropionin ≥95.0% (by GC)
Tripropionin
Formule:
C₁₂H₂₀O₆ MW: 260.29 g/mol Kookpunt: 176°C Vlampunt: 162°C |
MDL Nummer:
MFCD00027005 CAS nummer: 139-45-7 EINECS: 205-365-1 |
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...