Monoammonium glycyrrhizinate hydrate ≥75.0% (by HPLC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

G0151-1G G0151-25G
TCIAG0151-1GEA 17.6 EUR
TCIAG0151-1G TCIAG0151-25G
Monoammonium glycyrrhizinate hydrate ≥75.0% (by HPLC)
Monoammonium glycyrrhizinate hydrate
Formule: C₄₂H₆₅NO₁₆·xH₂O
Smeltpunt: 217°C
MDL Nummer: MFCD00167400
CAS nummer: 53956-04-0
EINECS: 258-887-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=646e35c65e8e0d3f474b52be5c456422456ca22c&vid=72fa5e7a32b9f79d7ad788346624966720a49792&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 75.0 area%
Specific rotation [a]20/D +45.0 to +52.0 deg (C=1.5, 40%EtOH) (calcd.on anh.substance)
Water max. 10.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR