Glyoxal 39% ∼8.8 mol/L in water

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

G0152-25G G0152-500G
TCIAG0152-25GEA 17.6 EUR
TCIAG0152-25G TCIAG0152-500G
Glyoxal 39% ∼8.8 mol/L in water
Glyoxal
Formule: C₂H₂O₂
MDL Nummer: MFCD00006957
CAS nummer: 107-22-2
EINECS: 203-474-9
Merck Index: 14,04509

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Concentration (Neutralization titration) 39.0 to 42.0 w/w%
Acid Value max. 10.0


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR