Copper(II) gluconate ≥97.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

G0275-25G G0275-100G
TCIAG0275-25GEA 17.6 EUR
TCIAG0275-25G TCIAG0275-100G
Copper(II) gluconate ≥97.0% (by titrimetric analysis)
Copper(II) gluconate
Formule: C₁₂H₂₂CuO₁₄
MDL Nummer: MFCD00075297
CAS nummer: 527-09-3
EINECS: 208-408-2
Merck Index: 14,02640

Order Now

Specification Test Results

Appearance Green to Blue powder to crystal
Purity (Chelometric Titration) min. 97.0 %
Specific rotation [a]20/D +14.0 to +19.0 deg (C=1, H2O)
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR