Glucan (from black yeast)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: beta-D-Glucan, Glucosan

G0331-1G G0331-5G
TCIAG0331-1GEA 74.8 EUR
TCIAG0331-1G TCIAG0331-5G
Glucan (from black yeast)
Glucan
Formule: (C₆H₁₀O₅)n
MDL Nummer: MFCD00466924
CAS nummer: 9012-72-0
EINECS: 232-739-1

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Beta-1,3-1,6-Glucan min. 80 %
Drying loss max. 8.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR