β-D-Glucopyranose-1-phosphate disodium salt ≥98.0% (by HPLC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

G0339-100MG G0339-20MG
TCIAG0339-100MGEA 1890 EUR
TCIAG0339-100MG TCIAG0339-20MG
β-D-Glucopyranose-1-phosphate disodium salt ≥98.0% (by HPLC)
beta-D-Glucopyranose-1-phosphate disodium salt
Formule: C₆H₁₃O₉P
Bewaar temperatuur: Vriezer
CAS nummer: 83833-15-2
EINECS: 280-989-5

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR