1-(2-Methoxyphenoxy)-2,3-epoxypropane ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: 1,2-Epoxy-3-(2-methoxyphenoxy)propane, Glycidyl Guaiacol ether, 2-[(2-Methoxyphenoxy)methyl]oxirane, Glycidyl 2-methoxyphenyl ether, Glycidyl 2-Methoxyphenyl ether

G0416-5G G0416-25G
TCIAG0416-5GEA 34.1 EUR
TCIAG0416-5G TCIAG0416-25G
1-(2-Methoxyphenoxy)-2,3-epoxypropane ≥98.0% (by GC)
1-(2-Methoxyphenoxy)-2,3-epoxypropane
Formule: C₁₀H₁₂O₃
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00190158
CAS nummer: 2210-74-4
EINECS: 218-644-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=690c900234e4289e686d7e4ef64e8b62b9378324&vid=690c900234e4289e686d7e4ef64e8b62b9378324&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to lump
Purity (GC) min. 98.0 %
Melting point 37.0 to 41.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR