Graphene, nanoplatelets, Length: 25 µm, Ø 6-8 nm

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

G0438-5G G0438-25G
TCIAG0438-5GEA 61.6 EUR
TCIAG0438-5G TCIAG0438-25G
Graphene, nanoplatelets, Length: 25 µm, Ø 6-8 nm
Graphene
MDL Nummer: MFCD00144065
CAS nummer: 1034343-98-0

Order Now

Specification Test Results

Appearance Black powder to crystal


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR