Graphene, nanoplatelets, Length: 15 µm, Ø 6-8 nm
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
G0442-5G
G0442-25G
TCIAG0442-5GEA
57.2
EUR
TCIAG0442-5G
TCIAG0442-25G
Graphene, nanoplatelets, Length: 15 µm, Ø 6-8 nm
Graphene
MDL Nummer:
MFCD00144065 CAS nummer: 1034343-98-0 |
Specification Test Results
Appearance | Black powder to crystal |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...