Glycoursodeoxycholic acid ≥95.0% (by HPLC, total nitrogen)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: N-(3alpha,7beta-Dihydroxy-5beta-cholan-24-oyl)glycine

G0459-200MG G0459-1G
TCIAG0459-200MGEA 56.1 EUR
TCIAG0459-200MG TCIAG0459-1G
Glycoursodeoxycholic acid ≥95.0% (by HPLC, total nitrogen)
Glycoursodeoxycholic acid
Formule: C₂₆H₄₃NO₅
MW: 449.63 g/mol
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD00137427
CAS nummer: 64480-66-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b6133292f3862f1f719e86265f516d139ee500a1&vid=b6133292f3862f1f719e86265f516d139ee500a1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (with Total Nitrogen) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR