1,4-Dioxane-2,5-dione ≥98.0% (by GC, titration analysis)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
G0467-5G G0467-25G
TCIAG0467-5GEA 30.8 EUR
TCIAG0467-5G TCIAG0467-25G
1,4-Dioxane-2,5-dione ≥98.0% (by GC, titration analysis)
1,4-Dioxane-2,5-dione
Formule: C₄H₄O₄
Kookpunt: 130°C
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD00081108
CAS nummer: 502-97-6
EINECS: 207-954-9
Order Now
Specification Test Results
Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization back titration) min. 98.0 %
Melting point 82.0 to 86.0 deg-C