Gastrodin ≥98.0% (by HPLC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

G0468-200MG G0468-1G
TCIAG0468-200MGEA 39.6 EUR
TCIAG0468-200MG TCIAG0468-1G
Gastrodin ≥98.0% (by HPLC)
Gastrodin
Formule: C₁₃H₁₈O₇
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00272169
CAS nummer: 62499-27-8
Merck Index: 14,04375

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 153.0 to 157.0 deg-C
Specific rotation [a]20/D -67.0 to -71.0 deg (C=1,H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR