Graphene, nanoplatelets aggregates, sub-micronparticles surface area 300 m²/g
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
G0500-5G
G0500-25G
TCIAG0500-5GEA
53.9
EUR
TCIAG0500-5G
TCIAG0500-25G
Graphene, nanoplatelets aggregates, sub-micronparticles surface area 300 m²/g
Graphene
MDL Nummer:
MFCD00144065 CAS nummer: 1034343-98-0 |
Specification Test Results
Appearance | Black powder to crystal |
Specific surface | 300.0 to 350.0 m2/g |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...