2-(N-Methylanilino)ethanol ≥97.0% (by titrimetric analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
N-Methyl-N-2-ethanolaniline, 2-(Methyl(phenyl)amino)ethanol, N-(2-Hydroxyethyl)-N-methylaniline
H0359-25ML
H0359-500ML
TCIAH0359-25MLEA
17.6
EUR
TCIAH0359-25ML
TCIAH0359-500ML
2-(N-Methylanilino)ethanol ≥97.0% (by titrimetric analysis)
2-(N-Methylanilino)ethanol
Formule:
C₉H₁₃NO MW: 151.21 g/mol Kookpunt: 266°C |
MDL Nummer:
MFCD00020572 CAS nummer: 93-90-3 EINECS: 202-285-9 |
Specification Test Results
Appearance | Light yellow to Brown clear liquid |
Purity (GC) | min. 95.0 % |
Purity (Nonaqueous Titration) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...