Heptyl sulphide ≥97.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
H0376-25G H0376-5G
TCIAH0376-25GEA 241 EUR
TCIAH0376-25G TCIAH0376-5G
Heptyl sulphide ≥97.0% (by GC)
Heptyl sulphide
Formule: C₁₄H₃₀S
Kookpunt: 298°C
Smeltpunt: –11°C
MDL Nummer: MFCD00027315
CAS nummer: 629-65-2
EINECS: 211-101-6
Order Now
Specification Test Results
Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %