Hexamethonium bromide ≥98.0% (by total nitrogen and titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

Synoniemen: N1,N1,N1,N6,N6,N6-hexamethylhexane-1,6-diaminium bromide, Hexane-1,6-bis(trimethylammonium Bromide), Hexamethylenebis(trimethylammonium Bromide)

H0481-25G H0481-500G
TCIAH0481-25GEA 48.4 EUR
TCIAH0481-25G TCIAH0481-500G
Hexamethonium bromide ≥98.0% (by total nitrogen and titration analysis)
Hexamethonium bromide
Formule: C₁₂H₃₀Br₂N₂
MW: 362.19 g/mol
MDL Nummer: MFCD00011787
CAS nummer: 55-97-0
EINECS: 200-249-7
Merck Index: 14,04688

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR