1-(3-Hydroxypropyl)-2-pyrrolidone ≥96.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: 1-(3-Hydroxypropyl)pyrrolidin-2-one
H0541-10ML H0541-25ML
TCIAH0541-10MLEA 86.9 EUR
TCIAH0541-10ML TCIAH0541-25ML
1-(3-Hydroxypropyl)-2-pyrrolidone ≥96.0% (by GC)
1-(3-Hydroxypropyl)-2-pyrrolidone
Formule: C₇H₁₃NO₂
MW: 143.19 g/mol
Kookpunt: 140°C
MDL Nummer: MFCD00059014
CAS nummer: 62012-15-1
EINECS: 263-373-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=28eb6a186abb839fabc2c825816adcc2d9e452da&vid=28eb6a186abb839fabc2c825816adcc2d9e452da&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Light yellow to Yellow to Orange clear liquid
Purity (GC) min. 96.0 %