Dimethyl itaconate ≥98.0% (by GC) stabilised
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Itaconic acid dimethyl ester, Methylenesuccinic acid dimethyl ester
Stabilisatie: Stabilised with hydroquinone (HQ)
I0206-500G I0206-25G
TCIAI0206-500GEA 83.6 EUR
TCIAI0206-500G TCIAI0206-25G
Dimethyl itaconate ≥98.0% (by GC) stabilised
Dimethyl itaconate
Formule: C₇H₁₀O₄
MW: 158.15 g/mol
Kookpunt: 109°C
Smeltpunt: 38
MDL Nummer: MFCD00008446
CAS nummer: 617-52-7
EINECS: 210-519-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=449af5f61ca2d1608e1eb79b7ca68a1565597bbe&vid=449af5f61ca2d1608e1eb79b7ca68a1565597bbe&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 98.0 %