2-(Carbobenzoxyamino)-1-ethanol ≥97.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: 2-(Cbz-amino)-1-ethanol, N-Z-Ethanolamine, Benzyl N-(2-Hydroxyethyl)carbamate, Benzyl (2-hydroxyethyl)carbamate, Z-Glycinol, N-(2-Hydroxyethyl)carbamic Acid Benzyl Ester

H0793-25G H0793-5G
TCIAH0793-25GEA 198 EUR
TCIAH0793-25G TCIAH0793-5G
2-(Carbobenzoxyamino)-1-ethanol ≥97.0% (by GC)
2-(Carbobenzoxyamino)-1-ethanol
Formule: C₁₀H₁₃NO₃
MDL Nummer: MFCD00191060
CAS nummer: 77987-49-6

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Melting point 58.0 to 64.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR