3-Hydroxyglutaronitrile ≥95.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

Synoniemen: 1,3-Dicyano-2-propanol

H0897-5G H0897-1G
TCIAH0897-5GEA 374 EUR
TCIAH0897-5G TCIAH0897-1G
3-Hydroxyglutaronitrile ≥95.0% (by GC)
3-Hydroxyglutaronitrile
Formule: C₅H₆N₂O
Kookpunt: 203°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00671545
CAS nummer: 13880-89-2
UN: 3276
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=77b38d1ed734d097887af81746400af35a435857&vid=77b38d1ed734d097887af81746400af35a435857&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Light yellow to Amber to Dark green clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR