8-Hydroxyjulolidine ≥97.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: 1,2,3,5,6,7-Hexahydropyrido[3,2,1-ij]quinolin-8-ol, 2,3,6,7-Tetrahydro-1H,5H-benzo[ij]quinolizin-8-ol

H0928-1G H0928-5G
TCIAH0928-1GEA 48.4 EUR
TCIAH0928-1G TCIAH0928-5G
8-Hydroxyjulolidine ≥97.0% (by HPLC, titration analysis)
8-Hydroxyjulolidine
Formule: C₁₂H₁₅NO
MDL Nummer: MFCD00006918
CAS nummer: 41175-50-2
EINECS: 255-247-9

Order Now

Specification Test Results

Appearance Light yellow to Brown powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 131.0 to 136.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR