Hexyl-7-(diethylamino)coumarin-3-carboxylate ≥98.0% (by HPLC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

H1145-1G H1145-5G
TCIAH1145-1GEA 51.7 EUR
TCIAH1145-1G TCIAH1145-5G
Hexyl-7-(diethylamino)coumarin-3-carboxylate ≥98.0% (by HPLC)
Hexyl-7-(diethylamino)coumarin-3-carboxylate
Formule: C₂₀H₂₇NO₄
MDL Nummer: MFCD06797113
CAS nummer: 851963-03-6

Order Now

Specification Test Results

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 65.0 to 69.0 deg-C

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR