2-[(Hydroxymethyl)amino]ethanol ≥95.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

H1174-25G H1174-5G
TCIAH1174-25GEA 79.2 EUR
TCIAH1174-25G TCIAH1174-5G
2-[(Hydroxymethyl)amino]ethanol ≥95.0% (by titrimetric analysis)
2-[(Hydroxymethyl)amino]ethanol
Formule: C₃H₉NO₂
CAS nummer: 65184-12-5
UN: 1760
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f88b37d4880618a7adf1801a7648d0390a2a916d&vid=f88b37d4880618a7adf1801a7648d0390a2a916d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Light yellow to Brown clear liquid
Purity (Neutralization titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR