(R)-9-(2-Hydroxypropyl)adenine ≥98.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: (R)-6-Amino-9-(2-hydroxypropyl)purine, (R)-1-(6-Amino-9H-purin-9-yl)propan-2-ol

H1291-1G H1291-5G
TCIAH1291-1GEA 17.6 EUR
TCIAH1291-1G TCIAH1291-5G
(R)-9-(2-Hydroxypropyl)adenine ≥98.0% (by HPLC, titration analysis)
(R)-9-(2-Hydroxypropyl)adenine
Formule: C₈H₁₁N₅O
MDL Nummer: MFCD07369451
CAS nummer: 14047-28-0

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 198.0 to 202.0 deg-C
Specific rotation [a]20/D -40.0 to -45.0 deg (C=1,EtOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR