Hexaphenylbenzene

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

H1412-1G H1412-5G
TCIAH1412-1GEA 73.7 EUR
TCIAH1412-1G TCIAH1412-5G
Hexaphenylbenzene
Hexaphenylbenzene
Formule: C₄₂H₃₀
Smeltpunt: 455°C
MDL Nummer: MFCD00003057
CAS nummer: 992-04-1
EINECS: 213-591-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe9eef6967746426d51976a52f8673e2cd77665e&vid=fe9eef6967746426d51976a52f8673e2cd77665e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Elemental analysis (Carbon) 92.40 to 97.20 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR