4-Hydroxy-3-methylbenzaldehyde ≥98.0% (by GC, titration analysis)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
H1419-1G H1419-5G
TCIAH1419-1GEA 102 EUR
TCIAH1419-1G TCIAH1419-5G
4-Hydroxy-3-methylbenzaldehyde ≥98.0% (by GC, titration analysis)
4-Hydroxy-3-methylbenzaldehyde
Formule: C₈H₈O₂
MW: 136.1479 g/mol
Smeltpunt: 118°C
MDL Nummer: MFCD00012360
CAS nummer: 15174-69-3
Order Now
Specification Test Results
Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 115.0 to 119.0 deg-C