1,1,3,3,5,5-Hexamethyltrisiloxane ≥97.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

Synoniemen: 1H,5H-Hexamethyltrisiloxane

H1441-5ML H1441-25ML
TCIAH1441-5MLEA 27.5 EUR
TCIAH1441-5ML TCIAH1441-25ML
1,1,3,3,5,5-Hexamethyltrisiloxane ≥97.0% (by GC)
1,1,3,3,5,5-Hexamethyltrisiloxane
Formule: C₆H₂₀O₂Si₃
Kookpunt: 128°C
Vlampunt: 20°C
MDL Nummer: MFCD00039788
CAS nummer: 1189-93-1
EINECS: 214-716-8
UN: 1993
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d3d88edcbb929f8648a77310c1f90838f2d3705f&vid=d3d88edcbb929f8648a77310c1f90838f2d3705f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR