Halobetasol Propionate ≥98.0% (by HPLC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

H1457-25MG H1457-100MG
TCIAH1457-25MGEA 42.9 EUR
TCIAH1457-25MG TCIAH1457-100MG
Halobetasol Propionate ≥98.0% (by HPLC)
Halobetasol Propionate
Formule: C₂₅H₃₁ClF₂O₅
Smeltpunt: 221°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00866013
CAS nummer: 66852-54-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6d087d0ecc578d3335de924e20fb0bc572f29043&vid=6d087d0ecc578d3335de924e20fb0bc572f29043&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Specific rotation [a]20/D +89.0 to +93.0 deg (C=1, Dioxane)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR