2-Hexyl-1-decanol ≥97.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
H1461-25ML H1461-100ML H1461-500ML
TCIAH1461-25MLEA 25.3 EUR
TCIAH1461-25ML TCIAH1461-100ML TCIAH1461-500ML
2-Hexyl-1-decanol ≥97.0% (by GC)
2-Hexyl-1-decanol
Formule: C₁₆H₃₄O
Kookpunt: 107°C
Smeltpunt: -15°C
Vlampunt: 159°C
MDL Nummer: MFCD00060903
CAS nummer: 2425-77-6
EINECS: 219-370-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=12dd165bf36e55424aac2b243e35aa87e1aec68b&vid=12dd165bf36e55424aac2b243e35aa87e1aec68b&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %