(4-(Hydroxymethyl)phenyl)acetic acid ≥97.0% (by titrimetric analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
2-(4-(Hydroxymethyl)phenyl)acetic acid, 4-(Hydroxymethyl)phenylacetic acid
H1473-1G
H1473-5G
TCIAH1473-1GEA
35.2
EUR
TCIAH1473-1G
TCIAH1473-5G
(4-(Hydroxymethyl)phenyl)acetic acid ≥97.0% (by titrimetric analysis)
(4-(Hydroxymethyl)phenyl)acetic acid
Formule:
C₉H₁₀O₃ MW: 166.18 g/mol |
MDL Nummer:
MFCD00065692 CAS nummer: 73401-74-8 |
|
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 94.0 % |
Purity (Neutralization titration) | min. 97.0 % |
Melting point | 128.0 to 132.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...