N-(2-Hydroxyethyl)maleimide ≥98.0% (by HPLC, total nitrogen)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
H1544-1G H1544-5G
TCIAH1544-1GEA 117 EUR
TCIAH1544-1G TCIAH1544-5G
N-(2-Hydroxyethyl)maleimide ≥98.0% (by HPLC, total nitrogen)
N-(2-Hydroxyethyl)maleimide
Formule: C₆H₇NO₃
Kookpunt: 130°C
MDL Nummer: MFCD00465266
CAS nummer: 1585-90-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4b62aa08b681678c3ffbb959d1d4df086078c21e&vid=4b62aa08b681678c3ffbb959d1d4df086078c21e&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 71.0 to 75.0 deg-C