4-(6-Hydroxyhexyloxy)benzoic acid ≥97.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
4-((6-Hydroxyhexyl)oxy)benzoic acid
H1629-1G
H1629-5G
TCIAH1629-1GEA
56.1
EUR
TCIAH1629-1G
TCIAH1629-5G
4-(6-Hydroxyhexyloxy)benzoic acid ≥97.0% (by GC)
4-(6-Hydroxyhexyloxy)benzoic acid
Formule:
C₁₃H₁₈O₄ |
MDL Nummer:
MFCD00971624 CAS nummer: 83883-25-4 |
|
Specification Test Results
Appearance | White to Almost white powder to crystaline |
Purity (GC) | min. 97.0 % |
Purity (Neutralization titration) | min. 96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...