2-Hexyl-1-n-octanol ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

H1635-1ML H1635-5ML
TCIAH1635-1MLEA 107 EUR
TCIAH1635-1ML TCIAH1635-5ML
2-Hexyl-1-n-octanol ≥98.0% (by GC)
2-Hexyl-1-n-octanol
Formule: C₁₄H₃₀O
Kookpunt: 162°C
MDL Nummer: MFCD00046769
CAS nummer: 19780-79-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=efc41049e3df2409198bcccb9cda6a53ab88334a&vid=efc41049e3df2409198bcccb9cda6a53ab88334a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR