1H,1H-Heptafluoro-n-butyl methacrylate ≥98.0% (by GC) stabilised
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
2,2,3,3,4,4,4-Heptafluorobutyl Methacrylate, 1H,1h-heptafluorobutylmethacrylate, Methacrylic acid 2,2,3,3,4,4,4-Heptafluorobutyl ester
Stabilisatie:
Stabilised with 4-methoxyphenol (MEHQ)
H1674-5G
H1674-25G
TCIAH1674-5GEA
52.8
EUR
TCIAH1674-5G
TCIAH1674-25G
1H,1H-Heptafluoro-n-butyl methacrylate ≥98.0% (by GC) stabilised
1H,1H-Heptafluoro-n-butyl methacrylate
Formule:
C₈H₇F₇O₂ MW: 268.13 g/mol Vlampunt: 37°C |
MDL Nummer:
MFCD00039251 CAS nummer: 13695-31-3 EINECS: 237-217-7 UN: 3272 ADR: 3 III |
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...