Hexazinone ≥98.0% (by HPLC, titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
H1686-5G
H1686-25G
TCIAH1686-5GEA
82.5
EUR
TCIAH1686-5G
TCIAH1686-25G
Hexazinone ≥98.0% (by HPLC, titration analysis)
Hexazinone
Formule:
C₁₂H₂₀N₄O₂ |
MDL Nummer:
MFCD00055526 CAS nummer: 51235-04-2 EINECS: 257-074-4 UN: 3077 ADR: 9,III Merck Index: 14,04699 |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 115.0 to 119.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...