Iminodiacetic acid disodium salt ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: Sodium 2,2'-azanediyldiacetate, Iminodiacetic acid disodium salt, Disodium Iminodiacetate hydrate

I0007-25G I0007-250G
TCIAI0007-25GEA 19.8 EUR
TCIAI0007-25G TCIAI0007-250G
Iminodiacetic acid disodium salt ≥98.0% (by titrimetric analysis)
Iminodiacetic acid disodium salt
Formule: C₄H₅NNa₂O₄
MDL Nummer: MFCD00043340
CAS nummer: 928-72-3
EINECS: 213-181-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=70e43a0a2293f5c66ca38586cc75d207e3679214&vid=70e43a0a2293f5c66ca38586cc75d207e3679214&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 % (calcd.on anh.substance)
Solubility in Water almost transparency
Water 8.0 to 17.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR