Diethanolamine hydrochloride ≥95.0% (by total nitrogen and titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

I0009-25G I0009-500G
TCIAI0009-25GEA 17.6 EUR
TCIAI0009-25G TCIAI0009-500G
Diethanolamine hydrochloride ≥95.0% (by total nitrogen and titration analysis)
Diethanolamine hydrochloride
Formule: C₄H₁₁NO₂
MDL Nummer: MFCD00012595
CAS nummer: 14426-21-2
EINECS: 238-396-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c5fadb5e63ec34fbe8761cb4f047aedb131c4c3f&vid=c5fadb5e63ec34fbe8761cb4f047aedb131c4c3f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Red to Green clear liquid
Purity (with Total Nitrogen) min. 95.0 %
Purity (Argentmetric Titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR