Allyl isonicotinate ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
Allyl 4-Pyridinecarboxylate, 4-Pyridinecarboxylic Acid Allyl Ester, Isonicotinic Acid Allyl Ester
I0136-25ML
I0136-5ML
TCIAI0136-25MLEA
354
EUR
TCIAI0136-25ML
TCIAI0136-5ML
Allyl isonicotinate ≥98.0% (by GC)
Allyl isonicotinate
Formule:
C₉H₉NO₂ Kookpunt: 106°C |
MDL Nummer:
MFCD00059786 CAS nummer: 25635-24-9 |
Specification Test Results
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...